missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl N-(3-thienyl)carbamate, 97%, Thermo Scientific™ Product Code.: 10120722

tert-Butyl N-(3-thienyl)carbamate, 97%, Thermo Scientific™

Product Code. 10120722
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10120722

Brand: Thermo Scientific Maybridge CC00092DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19228-91-2
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD01928808
InChI Key PRWYQCYSADTIBZ-UHFFFAOYSA-N
Synonym tert-butyl thiophen-3-ylcarbamate, tert-butyl n-3-thienyl carbamate, tert-butyl 3-thienylcarbamate, tert-butyl n-thiophen-3-yl carbamate, tert-butoxy-n-3-thienyl carboxamide, carbamic acid,n-3-thienyl-, 1,1-dimethylethyl ester, n-boc-3-aminothiophene, tert-butyl thien-3-ylcarbamate, 3-aminothiophene, n-boc protectedShow MoreShow Less
PubChem CID 736476
IUPAC Name tert-butyl N-thiophen-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CSC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-thienyl)carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-(3-thienyl)carbamate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.