Learn More
WTW™ Test Agent for OxiTop™ Systems
For use with Monitoring OxiTop™ systems; these test agents can be used for checking 6 measuring systems.
Brand: WTW™ 209333
589.42 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes calibration tablets
- For checking 6 measuring systems (long term test of leakage testing and negative pressure)
- For use with Monitoring OxiTop™ systems
- Test agent product type
Specifications
Test Agent | |
For checking 6 measuring systems (long term test of leakage testing and negative pressure) | |
Monitoring OxiTop™ systems | |
Calibration tablets |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- OxiTop PM
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid
Product Content Correction
Product Title
WTW™ Test Agent for OxiTop™ Systems > Includes: Calibration tablets; For use with: Checking 6 measuring systems
Spot an opportunity for improvement?Share a Content Correction