missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Test Tube Holder
Bochem™ Test Tube Holder for tubes with a diameter of 10 to 25mm.
Brand: Bochem™ 2282
This item is not returnable.
View return policy
Description
- For tube between 10 and 25mm diameter
Specifications
10 to 25mm diameter tubes | |
150 mm | |
Stainless Steel (18/10) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction