Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Brand:  Julabo™ 8970320
 View more versions of this product
Product Code. 11994698
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Test tube rack
EH-5, F12-ED, F12-EH, F12-MA, F25-ED open heating bath circulator
28 Test Tubes, 16/17 mm dia.
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Julabo™ Test Tube Racks for Open Heating Bath Circulator > Stainless Steel; Holds 28 Test tubes, Dia.: 16/17mm