Learn More
Texwipe™ TexBond™ 20 Cleanroom Bond Paper
Cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially formulated to address the needs of users requiring a paper that retains low ion and particle levels with significantly improved copying and writing qualities.
Brand:  Texwipe™ TX5719
 View more versions of this product
Product Code. 16405029
  • 149.00 DKK / Pack of 250
Only null left
Estimated Shipment: 14-08-2025
Description
  • Possesses both strength and excellent heat resistance, making it ideal for use in laser printers and photocopiers
  • Very low particle generation
  • Very low ion levels
  • High opacity rating
  • Precision cut edges
  • Cleanroom packaged Benefits
  • Exceptional copying ability
  • Laser printer and photocopier compatible
  • Clear reproductions
  • Autoclavable for use in sterile environments
  • Photocopying, ink jet and laser printing
  • Offset printing
  • Cleanroom manuals, work instructions, note taking and data transfer
  • Color coded carriers
Specifications
Specifications
White
210 x 297 mm
250 Sheets/Pk., 10 Packs/Cs.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Texwipe™ TexBond™ 20 Cleanroom Bond Paper > Color: White