Learn More
Texwipe™ ClipperMop™ Accessories and Replacements
Texwipe ClipperMop Accessories and Replacements, Polyethylene, Nylon, Tether Clips and Nylon Screws, For Use With: TX7102 and TX7103, highest quality cleanroom wipers, For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries, Recommended Applications: Safety
Brand: Texwipe™ TX7111B
3281.87 DKK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Texwipe ClipperMop Accessories and Replacements, Polyethylene, Nylon, Tether Clips and Nylon Screws, For Use With: TX7102 and TX7103, highest quality cleanroom wipers, For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries, Recommended Applications: Safety
- Clip Replacement kit
- Replacement parts for TX7102, TX7103
- There are 4 clips and 2 screws/pk.
Specifications
Replacement Pad | |
Clipper Mop Cleanroom Mop TX7103, Highest Quality Cleanroom Wipers | |
Foam |
White Fastners | |
For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
27Hours
11Minutes
4Seconds
32New Offer Save Up to 92%