Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Apply directly to floor
2265.00 DKK
Specifications
Quantity 60 Sheets/Mat
View More Specs
Products 1
Product Code Brand Color Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Color Dimensions (L x W) Price Quantity & Availability  
11575315
View Documents
Texwipe™
AMA254582W
White 45 x 25 in.
2265.00 DKK
Pack of 4
Only null left
Estimated Shipment: 05-08-2025
Add to basket
Description
Description
Apply directly to floor
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
60 Sheets/Mat
Documents
Product Certifications
Product Content Correction
Product Title
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer