missing translation for 'onlineSavingsMsg'
Learn More

TGR5/GPBAR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

2695.00 DKK - 3850.00 DKK

Specifications

Antigen TGR5/GPBAR1
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627397
View Documents
Novus Biologicals
NBP2-49376-25ul
25 ÎĽL
2695.00 DKK
25µL
Please to purchase this item. Need a web account? Register with us today!
18686577
View Documents
Novus Biologicals
NBP2-49376
0.1 mL
3850.00 DKK
0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TGR5/GPBAR1 Polyclonal antibody specifically detects TGR5/GPBAR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

TGR5/GPBAR1
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR
PBS (pH 7.2), 40% Glycerol
151306
IgG
Immunogen affinity purified
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR
This antibody was developed against a recombinant protein corresponding to amino acids: LGDQRYTAPWRAAAQRCLQGLWGRASRD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TGR5/GPBAR1 Antibody, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.