Learn More
Applied BioSystems™ Thermal Cycler Fleet Control Software
Stay in control with Thermal Cycler Fleet Control Software.
Brand:  Applied BioSystems™ A40070
Product Code. 16369666
  • 12490.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
Applied Biosystems™ Fleet Control Software is as intuitive and easy to use as our thermal cyclers. With this tool, you’ll be able to view and control all Applied Biosystems™ ProFlex™ PCR Systems, Automated Thermal Cyclers, and SimpliAmp™ Thermal Cyclers in your facility.
Secure—Fleet Control Software comes installed on a server that sits securely behind your firewall
Flexible—create custom permissions and rules for users, methods, and instruments
Powerful—quickly view your thermal cycler inventory and availability right from the dashboard
Convenient—control your thermal cyclers, reports, methods, and firmware upgrades from your office, another lab computer, or any other networked device
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/240 V
19.5 in. Wide-Viewing Angle
Thermal Cycler Fleet Control Software
1 each
Applied Biosystems™
Instrument Mangement and Control
PCR
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied BioSystems™ Thermal Cycler Fleet Control Software > Fleet Control Software comes pre-installed on Dell™ Optiplex™ 3050 all-in-one computer server with touchscreen, keyboard, and mouse
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.