Learn More
WTW™ Thermal Printer Paper for inoLab™ 7310P/9310P Printers
High quality thermal printer paper for use with inoLab™ 7310/9310 printers.
Brand:  WTW™ 250115
133.55 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15239885
  • 152.00 DKK / Each
Only null left
Estimated Shipment: 30-06-2025
Description
  • Thermal printer paper
  • For use with inoLab™ 7310/9310 printers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermal Printer Paper
9 m
inoLab 7310/9310
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Thermal Printer Paper for inoLab™ 7310P/9310P Printers > Width: 58mm; For use with: inoLab 7310/9310