Learn More
Brady™ Thermal Transfer Printer Labels: Rectangular
Rectangular printable labels for thermal transfer printers.
Brand:  Brady™ 121010
 View more versions of this product
Product Code. 17140161
  • 1710.00 DKK / Pack of 2000
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
Printer Compatibility
  • BBP81
  • Brady 1244
  • Brady 1344
  • Brady 200MVP Plus
  • Brady 2461
  • Brady 300MVP Plus
  • Brady 300X-Plus II
  • Brady 3481
  • Brady 360X-Plus II
  • Brady 600X-Plus II
  • Brady 6441
  • Brady IP
  • Tagus T200
  • Tagus T300
  • Thermal Transfer Printers
This material is UL recognized with its respective ribbon; use Brady Ribbon Part# R4902.
Order Info
See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications
Specifications
Acrylic
Thermal Transfer Printer Label
3 in. Core
White
2000/Roll
25.4 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Thermal Transfer Printer Labels: Rectangular > Size: 25.4W x 25.4mm H