Learn More
Texwipe™ ThermaSeal™ 70 Wipers
Offers quality with superior absorbency.
Brand:  Texwipe™ TX2074
Product Code. 15752396
  • 805.00 DKK / Pack of 600
Only null left
Estimated Shipment: 06-08-2025
Description
  • Hot-cut polyester wipers for all-purpose cleaning in critical cleanroom environments
  • They offer high absorbency, low particles and low ions
  • Produced and packaged in a state-of-the-art ISO Class 4 cleanroom environment
  • Features 7 thermally sealed edges for advanced particle and fiber control
  • Laundered and packaged in an ISO Class 4 cleanroom environment
  • 100% continuous-filament,double-knit polyester fabric
  • Solvent-safe Bag-Within-A-Bag™ cleanroom packaging
  • Superior absorbency, wiping efficiency and abrasion resistance
  • Designed for ISO Class 3-5 cleanroom applications
  • Ideal for wiping interior of process tools and medical devices
Specifications
Specifications
10 x 10 cm
600 Wipers/ Bag (2 inner Bags of 300 Wipes), 10 Bags/ Cs.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Texwipe™ ThermaSeal™ 70 Wipers > Dimensions (L x W): 10 x 10 cm