Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Brand:  Thermo Scientific™ 01092
1334.79 DKK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
 View more versions of this product
Product Code. 15129485
  • 1732.10 DKK / Each
Illuminating Protein Pathways
Use promo code 24148 at checkout
View Available Promotions
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 Ă— 500mL (bottles), 6 Ă— 550mL (blood bags), and 132 Ă— 7mL (tubes). Max. O.D. Ă— L: 70 x 160mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 x 250 mL Bottles
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ H-6000A Bucket Adapters > 250mL bottle