Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Brand: Thermo Scientific™ 260887
3588.71 DKK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Description
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Includes Barcode | |
Standard | |
96F well plate, Non-Treated, clear, with lid, Sterile, Barcoded (Pack of 10, Case of 160) | |
10 Pk. | |
Clear | |
High Flange | |
400 ÎĽL | |
Sterile | |
Flat Bottom (F-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
With Lid | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Non-treated | |
160 | |
160/Cs. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
186518 | Certificate of Compliance | 27/06/2025 | 260887 |
186025 | Certificate of Compliance | 21/03/2025 | 260887 |
185546 | Certificate of Compliance | 12/02/2025 | 260887 |
185546 | Certificate of Irradiation | 09/05/2025 | 260887 |
184072 | Certificate of Irradiation | 07/02/2025 | 260887 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.