Learn More
Thermo Scientific™ Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
Collection plates and balance plates for use with 96-well filter spin plates
Brand: Thermo Scientific™ 89813
427.17 DKK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Specifications
0.0 | |
Collection Plate | |
96 | |
6 plates |
Pierce | |
Polypropylene | |
96-Well Plates |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Invitrogen
,
ABI Applied Biosystems
,
Thermo Scientific
,
Affymetrix eBioscience
Nurture Your Progress
Exclusive savings to help your research thrive.Promo Code: 24108Expires: 2025-06-27
Days
17Hours
6Minutes
14Seconds
44New Offer Save Now -
Multiple Manufacturers
Get 3 for 2
Exclusive savings, limited-time offer.Promo Code: 24112Expires: 2025-06-26
Days
16Hours
6Minutes
14Seconds
44New Offer Save Now