Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Brand: VEE GEE 80901E
Description
- FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial
Specifications
Safety Blue | |
Partial Immersion | |
Glass | |
Safety Blue | |
Serialized Glass Thermometer | |
48.9°C | |
-34.4°C | |
-34.4 to +48.9°C | |
1.1°C (2°F) | |
1.5 cm |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
30.48 cm | |
Immersion/General Purpose (Tapered) | |
5 g | |
120°F | |
-30°F | |
-30 to +120°F | |
±1 subdivision | |
To immersion line |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.