missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Brand: VEE GEE 80704E
This item is not returnable.
View return policy
Description
- FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial
Specifications
Safety Blue | |
Total Immersion | |
Glass | |
Safety Blue | |
Serialized Glass Thermometer | |
104.4°C | |
-1.1°C | |
-1.1 to 104.4°C | |
1.1°C (2°F) | |
1.52 cm |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
39 cm | |
Immersion/General Purpose (Tapered) | |
5 g | |
220°F | |
30°F | |
30 to 220°F | |
±1 subdivision | |
39 cm (15.4 in.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VEE GEE Thermometer, Encapsulated in FEP > Immersion Length: 39 cm (15.4 in.), Immersion Type: Total Immersion, Temperature Range: 30 to 220°F
Spot an opportunity for improvement?Share a Content Correction