Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Brand:  VEE GEE 80908E
 View more versions of this product
Product Code. 17881214
  • 530.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
  • Feature Safety Blue fill against a white background for easy reading
  • Accuracy is equal to one subdivision
  • Immersion: Total or Partial
Specifications
Specifications
Safety Blue
Partial Immersion
Glass
Safety Blue
Serialized Glass Thermometer
150°C
-20°C
-20 to 150°C
1°C (1.8°F)
1.5 cm
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
30.48 cm
Immersion/General Purpose (Tapered)
5 g
302°F
-4°F
-4 to +302°F
±1 subdivision
To immersion line
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VEE GEE Thermometer, Encapsulated in FEP > Immersion Length: To immersion line, Immersion Type: Partial Immersion, Temperature Range: -4 to +302°F