Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand:  SymLine™ 106523
696.73 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
 View more versions of this product
Product Code. 15886024
  • 865.00 DKK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 25-08-2025
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
GL25 (f) to GL25 (f)
Black
HDPE-EC
Thread Adapter
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number57277UN25 Certificate TypeCertificate of Analysis Date27/06/2025 Catalog Number
Lot Number42699UN25 Certificate TypeCertificate of Analysis Date14/06/2025 Catalog Number
Lot Number27007UN25 Certificate TypeCertificate of Analysis Date15/03/2025 Catalog Number106523
Lot Number07024UN25 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog Number106523
Promotions
Promotions
  • Multiple Manufacturers
    From wipes to waste bins, labels to liners – save on them all.
    Promo Code: 24132
    Expires2025-08-24
    New Offer
    Shop Now
Product Content Correction
Product Title
SymLine™ Adapters > Adapters; Description: Thread Adapter, GL25 (f) to GL25 (f); Material: HDPE-EC; Type: Adapter