Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
287.00 DKK - 1035.00 DKK
Specifications
Product Type Thread Adapter
View More Specs
Products 31
Product Code Brand Closure Size Closure Color Price Quantity & Availability  
Product Code Brand Closure Size Closure Color Price Quantity & Availability  
10800391
View Documents
SCAT™
108021
Thread 1: 2 in. BSP/G2 in. (f), Thread 2: S60 (m) Gray
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10890381
View Documents
SCAT™
107091
Thread 1: 63 B (f), Thread 2: S60/61 (m) White
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10795821
View Documents
SCAT™
107994
Thread 1: GL/S40 (f), Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10151260
View Documents
SCAT™
107995
Thread 1: GL38, Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10600051
View Documents Promotion Details
SCAT™
107094
Thread 1: GL45 (f), Thread 2: S55 (m) Colorless
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
14716301
View Documents
SCAT™
108081
Thread 1: GL45 (m), Thread 2: S40/41 (f) Colorless
478.00 DKK
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
10859680
View Documents
SCAT™
107015
Thread 1: GL45 (m), Thread 2: 2 in. Tri-Sure (m) White
478.00 DKK
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10384900
View Documents
SCAT™
107996
Thread 1: GL45 (m), Thread 2: GL32 (f) Colorless
478.00 DKK
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
10620061
View Documents
SCAT™
107014
Thread 1: GL45 (m), Thread 2: R2 in. BSP/G2 in. (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10364900
View Documents
SCAT™
107085
Thread 1: GL45, Thread 2: R1 1/2 in. (m) White
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10160050
View Documents
SCAT™
107022
Thread 1: R2 in. BSP/G2 in. (f), Thread 2: 2 in. Trisure (m) Orange
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10661001
View Documents
SCAT™
108022
Thread 1: S60 (m), Thread 2: R2 in. BSP/G2 in. (m) Black
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10130580
View Documents
SCAT™
107028
Thread 1: R 1 1/2 in. (f), Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10809350
View Documents
SCAT™
107024
Thread 1: R2 in. BSP/G2 in. (f), Thread 2: S55 (f) Green
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10314370
View Documents
SCAT™
107023
Thread 1: R2 in. BSP/G2 in. (f), Thread 2: S60/61 (f) Yellow
287.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10130180
View Documents Promotion Details
SCAT™
107093
Thread 1: S51 (f), Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10508130
View Documents
SCAT™
107099
Thread 1: S55 (f), Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
10211750
View Documents Promotion Details
SCAT™
107090
Thread 1: S 60/61 (f) Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10323870
View Documents
SCAT™
107089
Thread 1: S60/61, Thread 2: S55 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10347759
View Documents Promotion Details
SCAT™
108061
Thread 1: S50 (f), Thread 2: GL45 (m) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10170130
View Documents
SCAT™
107092
Thread 1: S51 (m), Thread S55 (f) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10671991
View Documents
SCAT™
107086
Thread 1: S51 (m), Thread 2: S60/61 (f) Colorless
478.00 DKK
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10292480
View Documents
SCAT™
107084
Thread 1: S55 (m), Thread 2: S50 (f) Colorless
625.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10580061
View Documents
SCAT™
107074
Thread 1: S60/61 (f), Thread 2: GL25 (m) Colorless
670.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10755372
View Documents
SCAT™
108029
Thread 1: S60/61 (m), Thread 2: 2 in. Tri-Sure (m) Colorless
670.00 DKK
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10598120
View Documents
SCAT™
107097
Thread 1: S60/61 (m), Thread 2: S51 (f) Colorless
625.00 DKK
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10150610
View Documents
SCAT™
107096
Thread 1: S60/61 (m), Thread 2: S55 (f) Colorless
478.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10670451
View Documents
SCAT™
107079
Thread 1: S71 (f), Thread 2: GL45 (m) Colorless
865.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17893520
View Documents Promotion Details
SCAT™
108279
Thread 1: S60/61 (m), Thread 2: GL45 (f) Colorless
670.00 DKK
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
17987240
View Documents
SCAT™
107146
Thread 1: GL45 (m), Thread 2: S65 (f) Gray
664.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17927270
View Documents
SCAT™
108066
Thread 1: GL45 (m), Thread 2: B83 (f) Gray
1035.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
Thread Adapter
Documents
Product Certifications
Product Content Correction
Product Title
SCAT™ Polypropylene Thread Adapters