Learn More
Amarell™ Electronic Timer-Clock
Includes an alarm and stopwatch functionality. The electronic timer-clock will countdown from or up to 24 hours.
Brand:  Amarell™ E920630
Product Code. 10329860
  • 136.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
  • Countdown or count-up timer with loud alarm signal
  • With clock and stopwatch memory function
  • Up to 24 hours
Specifications
Specifications
Timer
Yes
68 x 53 x 20 mm
24 hr.
LR44/A76
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Amarell™ Electronic Timer-Clock > Dimensions: 68 x 53 x 20mm