Learn More
Timosaponin AIII, MedChemExpress
MedChemExpress Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Brand:  Medchem Express HY-N0810
Encompass_Preferred
 View more versions of this product
Product Code. 16441518
  • 795.16 DKK / 5mg
Explore more special offers
Description
Description
Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Specifications
Specifications
Timosaponin AIII
White
C39H64O13
DMSO : 125 mg/mL (168.71 mM; Need ultrasonic)
740.92
98.0%
Solid
41059-79-4
Research
5 mg
C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@H](C)CO4)([H])C[C@@]1([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC2
740.92
Research
Cancer-programmed cell death