missing translation for 'onlineSavingsMsg'
Learn More
Tin Standard For ICP, 1000μg/mL in 20% HCl, Assurance Grade, SPEX CertiPrep™ Product Code.: 10651563
compliance-icons compliance-icons

Tin Standard For ICP, 1000μg/mL in 20% HCl, Assurance Grade, SPEX CertiPrep™

Product Code. 10651563 Shop All SPEX Certiprep Products
500 mL, Triple-leached LDPE bottle
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy

Product Code. 10651563

Brand: SPEX Certiprep PLSN52X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sn1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-31-5, 7647-01-0, 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 1.0, 20.0, 79.0
Quantity 500 mL
CAS Max % 1.0, 20.0, 79.0
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 20% HCl
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
1% HNO3/1% HF    1000μg/mL    PLSN2-2X
2% HNO3/2% HF    10,000    PLSN2-3X
20% HCl    1000μg/mL    PLSN5-2Y, -2T, -2X    10,000    PLSN5-3Y, -3X
Chemical Name or Material Tin Standard
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P364-And wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin Standard For ICP, 1000μg/mL in 20% HCl, Assurance Grade, SPEX CertiPrep™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.