Learn More
Lovibond™ Tintometer™ Sulphite No.2 L.R.Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505331
This item is not returnable.
View return policy
Description
- Sulphite No.2 L.R.Tablets
- Bottle of 250
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
250 Tablets | |
Sulphite No.2 L.R.Tablets |