Learn More
Kartell™ Tissue Embedding Cassettes - Uni Type
For paraffin inclusions, Universal type, provided with clipon closure lid that can easily be removed
Brand:  Kartell™ 0292110
Additional Details : Weight : 5.54000kg
Product Code. 15937860
  • 1760.00 DKK / Pack of 2000
Estimated Shipment: 22-07-2024
to see stock.
Description
Description
  • CE marked, IVD ceritfied
  • Universal Type
  • Clipon closure lid that can easily be removed
  • Specifications
    Specifications
    500
    40 mm
    Polyoxymethylene (POM)
    Red
    28 mm