Learn More
SI Analytics TitroLine™ TL 7000-M2/20
Features the highest safety for measuring and calibrating through wireless sensor recognition for ID-electrodes. SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M2/20 possesses general titration feautres but offers improved performace.
Brand: SI Analytics 285220170
44898.21 DKK valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- The brilliant TFT display allows perfect readout even from the side
- The intelligent exchangeable units are storing all relevant reagent and unit data
- The automatic wireless recognition of SCHOTT™ Instruments
- ID electrodes guarantees accurate calibration and measurements
- 3 USB and 2 RS-232 ports allow to connenct e.g. USB keyboards, USB printers, USB storage media, stirrer, balance, PC and other
- SI Analytics instruments
Specifications
TitroLine 7000 Basic Unit | |
Polypropylene | |
10°C to 40°C | |
15.3 cm |
Brown Glass Bottle For Titrant, GL 45 And S 40-Bottle Adapter, Hoses, Drip Off Tube And Titration Tip. With pH-Combination Electrode IL-pHT-A120MF-DIN-N And Buffer Set | |
TL 7000-M2/20 | |
45 cm | |
29.6 cm |