Learn More
TMEM41B Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-12857FR
Product Code. 18310887
  • 4695.34 DKK / 0.10mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
TMEM41B
Polyclonal
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Affinity-purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
DyLight 680
KIAA0033MGC33897, MGC57262, transmembrane protein 41B
Anti-TMEM41B antibody was raised against a peptide corresponding to 10 amino acids near the carboxyl terminus of human TMEM41B. The immunogen is located within the last 50 amino acids of TMEM41B.
0.1 mL
Autophagy, Lipid and Metabolism
440026
Store at 4°C in the dark.
IgG