Learn More
TN buffer soln., Thermo Scientific™ Product Code.: 15426129
TN buffer soln., Thermo Scientific™
Product Code. 15426129
500 mL, 500mL
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Product Code. 15426129
Brand: Thermo Scientific Alfa Aesar J63985.AP
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TN buffer soln.
Color Colorless
Physical Form Liquid
Quantity 500 mL
Synonym TRIS NaCl buffer soln
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Product Code
Lot NumberP28E519 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberF21X565 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot NumberF21X564 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction
Product Title
TN buffer soln., Thermo Scientific™ >