Learn More
Abnova™ TNFRSF13C 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Brand:  Abnova™ H00115650-T01.100uL
Product Code. 16167233
  • 1710.00 DKK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
293T
TNFRSF13C 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 20.35kDa
Transient overexpression cell lysate was tested with anti-TNFRSF13C antibody (Mfr. No. H00115650-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_052945.2
pCMV-TNFRSF13C full-length
100 μL
Human