Learn More
TNP-ATP triethylammonium salt, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
Description
High affinity, selective P2X receptor antagonist. Inhibits ATP-induced currents in cells expressing P2X1, P2X3 and heteromeric P2X2/3 receptors with IC50 values of 6, 0.9 and 7 nM respectively. Displays 1000-fold selectivity over P2X2, P2X4 and P2X7.
Specifications
Specifications
CAS | 61368-63-6 |
Molecular Formula | C16H13N8O19P3·C24H64N |
Quantity | 5 mg |
Synonym | 2',3'-O-(2,4,6-Trinitrophenyl)adenosine-5'-triphosphate tetra(triethylammonium) salt |
Formula Weight | 1123.04 |
Percent Purity | >95% |
Chemical Name or Material | TNP-ATP triethylammonium salt |