Learn More
Tocris Bioscience™ Caffeic acid-pYEEIE
Phosphopeptide ligand for src SH2 domain
Brand: Tocris Bioscience™ 1935/1
1232.38 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Caffeic acid-pYEEIE is a phosphopeptide ligand for the src SH2 domain (IC50 = 42 nM); displays 30-fold higher affinity than N-acetyl-O-phosphono-Tyr-Glu-Glu-Ile-Glu ( Ac-pYEEIE, Cat. No. 1927).Specifications
1 mg |