missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tomosyn Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1485.92 DKK - 3498.83 DKK
Specifications
Antigen | Tomosyn |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310537
|
Bio-Techne
NBP3-03781-20UL |
20 μL |
1485.92 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331456
|
Bio-Techne
NBP3-03781-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tomosyn Polyclonal specifically detects Tomosyn in Human samples. It is validated for Western Blot.Specifications
Tomosyn | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
134957 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
FLJ30922, Lethal(2) giant larvae protein homolog 3, LGL3, LLGL3Nbla04300, MGC141942, MGC141968, putative protein product of Nbla04300, syntaxin binding protein 5 (tomosyn), syntaxin-binding protein 5, tomosyn, tomosyn-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 640-900 of human Tomosyn (NP_640337.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |