Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Top Position Access Port is an accessory for use on the top of a variety of chambers and incubators.
Brand:  BINDER™ 8012-0386
2954.54 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15631726
  • 3250.00 DKK / Each
Only null left
Estimated Shipment: 03-09-2025
Description
  • For use on top
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 10 mm, top
BINDER Chambers Series BD
Top Position
10 mm
Access Port
Silicone Plug
0.4 in.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Access Port with Silicone Plug > Diameter Metric: 10 mm, For Use With: BINDER Chambers Series BD