Learn More
Unity Lab Services™ Total Care Warranty for Thermo Scientific CO2 Incubators
Total Care Warranty provides an upgrade to factory warranty. Includes 2 day response time (within specified location and criteria), priority technical support, and one preventative maintenance visit.
Brand:  Unity Lab Services™ TCWTYINCCO2
4199.85 DKK valid until 2025-06-30
Use promo code "24077" to get your promotional price.
Product Code. 17250404
  • 4941.00 DKK / Each
Exclusive Promotion on Equipment and Instruments
Use promo code 24069 at checkout
View Available Promotions
Only null left
Estimated Shipment: 24-10-2025
Description
  • Upgrade a basic factory warranty with priority Technical Support response, and faster on-site scheduling along with an annual preventative maintenance visit
  • 2 business day response time (may vary by location) for corrective maintenance
  • Includes parts and labor covered under factory warranty for corrective maintenance
  • One scheduled Preventative Maintenance visit, including a preventative maintenance kit (where applicable)
  • Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools.
  • Access to online service knowledge base
  • Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the terms and conditions
  • Instrument Software/ Firmware updates included as needed to support a preventative maintenance or corrective maintenance visit
  • Upgrades to new versions of software are not included
  • 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CO2 Incubators
Priority Technical Support, and One Preventative Maintenance Visit
Total Care Warranty
Upgrades to New Software Versions
Upgrade to Factory Warranty
2 Day Response Time (Within Specified Location and Criteria), Priority Technical Support, and One Preventative Maintenance Visit
2 Days (Within Specified Location and Criteria)
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Take advantage of our limited-time offer on top-quality equipment and instruments to elevate your research and laboratory capabilities.
    Promo Code: 24069
    Expires2025-06-30
    Days
    20
    Hours
    17
    Minutes
    10
    Seconds
    25
  • ULS Unity Lab Services , Thermo Scientific
    Save now on our latest CO2 incubator and ULT freezer, plus services.
    Promo Code: 24077
    Expires2025-06-30
    Days
    20
    Hours
    17
    Minutes
    10
    Seconds
    25
    New Offer
    Shop Now
Product Content Correction
Product Title
Unity Lab Services™ Total Care Warranty for Thermo Scientific CO2 Incubators > CO2 Incubators