Learn More
WTW™ Total Organic Carbon Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 252036
3170.91 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description

Specifications
For measurement of TOC (Total organic carbon) | |
Certificates | |
5 to 80 mg/L | |
Photometric |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
Sodium (Na) cell test kit |
Safety and Handling
- TOC Cell Test Method: photometric 5.0 - 80.0 mg/l Spectroquant
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.