Learn More
TPO agonist 1, MedChemExpress
MedChemExpress TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia.

1248.83 DKK - 14985.99 DKK
Specifications
Chemical Name or Material | TPO agonist 1 |
---|---|
CAS | 1033040-23-1 |
Color | Red |
Purity Grade Notes | Research |
Molecular Formula | C25H22N8O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16475717
![]() |
Medchem Express
HY-100380 |
5 mg |
1752.39 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16459546
![]() |
Medchem Express
HY-100380 |
50 mg |
9366.25 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16452978
![]() |
Medchem Express
HY-100380 |
10 mg |
2749.45 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434296
![]() |
Medchem Express
HY-100380 |
100 mg |
14985.99 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16400016
![]() |
Medchem Express
HY-100380 |
2 mg |
1248.83 DKK
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia.Specifications
TPO agonist 1 | |
Red | |
C25H22N8O2 | |
O=C(N(C1=CC=C(C)C(C)=C1)N=C/2C)C2=N\NC3=C(O)C(C4=CC=CC(C5=NN=NN5)=C4)=CC=C3 | |
466.49 | |
Research | |
COVID-19-immunoregulation |
1033040-23-1 | |
Research | |
DMSO : ≥ 36 mg/mL (77.17 mM) | |
466.49 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.