Learn More
Thermo Scientific™ TRACE™ GC Columns for Pesticides
Identify organophosphate and organochlorine pesticides with high performance, sensitivity and reproducibility.
Brand: Thermo Scientific™ 26RC142P
4304.45 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Identify organophosphate and organochlorine pesticides with high performance, sensitivity, and reproducibility using the Thermo Scientific™ TRACE™ GC Columns for Pesticides. These columns are specifically designed and tested to meet the most stringent requirements for the analysis of pesticides and herbicides.
Excellent Quality and Reproducibility:- Low bleed decreases MS contamination
- Innovative design facilitates use at higher temperatures
- Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
- Chlorinated aromatics
- Plant sterols
- Pyrethroids
Specifications
280/300°C | |
0.25 mm | |
Trace™ Pesticide IV | |
Pesticides Analysis | |
30 m |
0.25 mm | |
TRACE | |
G43 | |
0.25 μm | |
30 m x 0.25 mm x 0.25 μm |