Learn More
BRAND™ Transferpette™ S Pipette Wall Holder
Accessory for use with Transferpette™ S single and multichannel pipettes.
Brand: BRAND™ 704812
184.30 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Allows handing of pipette on walls
- Pipettes sold separately
Specifications
Wall Mount | |
Transferpette™ S Pipettes |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%