Learn More
Transketolase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11039-1-AP-150UL
 View more versions of this product
Product Code. 16817843
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
Specifications
Transketolase
Polyclonal
Unconjugated
TKT
TK, TKT, TKT1, transketolase
Rabbit
Antigen Affinity Chromatography
RUO
21881, 64524, 7086
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29401, P40142, P50137
Tkt
Transketolase Fusion Protein Ag1519
150 ÎĽL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Transketolase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated