Learn More
Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Providing a number of advantages over conventional peroxide cured silicones. Fisherbrand™ Translucent Platinum Cured Silicone Tubing is available in a comprehensive range of stock sizes.
Brand:  Fisherbrand™ 1170-8178
 View more versions of this product
Product Code. 11708178
  • 1645.00 DKK / Each
Only null left
Estimated Shipment: 23-06-2025
Description
  • Autoclavable
  • Non-yellowing
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • No peroxide byproducts
  • High tear strength as standard
  • Extended life in Peristaltic pumps
  • Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Specifications
FDA, BFR and WRAS approved
16 mm
20.8 mm
2.4 mm
-40°C
Flexible
200°C
Autoclavable
Platinum-cured Silicone
25 m
Ideal for biological, pharmaceutical, biotechnology, aerospace, hot work
Translucent
0.5 bar
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Translucent Platinum Cured Silicone Tubing > I.D.: 16mm; O.D.: 20.8mm; Length: 25m; Nominal pressure: 0.5bar