Learn More
BRAND™ Translucent Polypropylene T-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with T-style side arm.
Brand:  BRAND™ 152810
 View more versions of this product
Product Code. 11813930
  • 224.00 DKK / Pack of 20
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Description
  • Translucent polypropylene construction
  • Accommodates 3 tubes of the same diameter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Connector
55 mm
20 Pack
T-shape
Translucent
Polypropylene
8 to 9 mm tubing I.D.
8 to 9 mm tubing I.D.
39 mm side arm