Learn More
Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap
Brand:  Kartell™ 0143200
Product Code. 10338561
  • 1755.00 DKK / Pack of 20
Only null left
Estimated Shipment: 30-07-2025
Description
These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap > Capacity: 100mL