Learn More
Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap
Brand: Kartell™ 0143200
This item is not returnable.
View return policy
Description
These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap > Capacity: 100mL
Spot an opportunity for improvement?Share a Content Correction