Learn More
Fisherbrand™ Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Brand: Fisherbrand™ 1174-3845
This item is not returnable.
View return policy
Description
- Translucent silicone tubing
- FDA compliant (food)
Specifications
FDA, BFR and WRAS approved | |
1 mm | |
3 mm | |
1 mm | |
Pharmaceutical , biotechnology, transfer sterile fluids | |
Sterile | |
Flexible |
Autoclavable | |
Silicone | |
5 m | |
Peristaltic pumps | |
-40°C | |
Translucent | |
250°C |
Product Content Correction
Product Title
Fisherbrand™ Translucent Silicone Tubing > I.D.: 1mm; O.D.: 3mm; Length: 5m
Spot an opportunity for improvement?Share a Content Correction