Learn More
Fisherbrand™ Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Brand:  Fisherbrand™ 1178-3845
 View more versions of this product
Product Code. 11783845
  • 102.00 DKK / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Description
  • Translucent silicone tubing
  • FDA compliant (food)
Specifications
Specifications
FDA, BFR and WRAS approved
Silicone
5 m
Peristaltic pumps
-40°C
Translucent
250°C
3 mm
7 mm
2 mm
Pharmaceutical , biotechnology, transfer sterile fluids
Sterile
Flexible
Product Title
Fisherbrand™ Translucent Silicone Tubing > I.D.: 3mm; O.D.: 7mm; Length: 5m