missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
Description
- Polypropylene spray tube
- With coloured VENT-CAP for easy visual identification
- Safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS):
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Practically no leakage or dripping with the VENT-CAP
Specifications
Specifications
Capacity (Metric) | 250 mL |
Material | Polypropylene |
Thread Size (Metric) | GL 45 |
Closure Color | Red |
Mouth | Wide |
Height (Metric) | 146 mm |
Product Type | Safety Wash Bottle |
Diameter (Metric) | 58 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VITLAB™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles >
Spot an opportunity for improvement?Share a Content Correction