Learn More
Bochem™ Transport Basket
Bochem™ Transport Basket is a stackable metal mesh basket with handles.
Brand: Bochem™ 17302
This item is not returnable.
View return policy
Description
- 10 x 10 x 3mm mesh dimensions
- Stainless Steel
Specifications
EURO-STANDARD | |
Stackable | |
18/10 E-Poli | |
300 x 400 x 150mm |