Learn More
Tri-n-butylphosphine, 95%, AcroSeal™ Product Code.: 10402324
Tri-n-butylphosphine, 95%, AcroSeal™
Product Code. 10402324 Shop All Thermo Scientific Chemicals Products
100 mL, AcroSeal™ Glass bottle, 100mL
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
Product Code. 10402324
Brand: Thermo Scientific Acros 427301000
Request Bulk or Custom Format
CAS: 998-40-3 | C12H27P | 202.32 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 998-40-3
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00009462
InChI Key TUQOTMZNTHZOKS-UHFFFAOYSA-N
Synonym tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbpShow MoreShow Less
PubChem CID 13831
IUPAC Name tributylphosphane
SMILES CCCCP(CCCC)CCCC
Specifications
Melting Point -65.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 240.0°C to 242.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula [CH3(CH2)3]3P
Refractive Index 1.4610 to 1.4630
Quantity 100 mL
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with ether,methanol,ethanol and benzene
Formula Weight 202.32
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-butylphosphine
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0452611 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code427308000,427301000
Lot NumberA0467247 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code427308000,427301000
Lot NumberA0467247 Certificate TypeCertificate of Analysis Date28/10/2024 Product Code427308000,427301000
Lot NumberA0465396 Certificate TypeCertificate of Analysis Date09/08/2024 Product Code427308000,427301000
Lot NumberA0456185 Certificate TypeCertificate of Analysis Date31/03/2024 Product Code427308000,427301000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H250-Catches fire spontaneously if exposed to air.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Product Title
Tri-n-butylphosphine, 95%, AcroSeal™ >