missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, 20% w/v, Extra Pure, Scharlab
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.
Specifications
Specifications
CAS | 76-03-9 |
Molecular Formula | C2HCl3O2 |
Linear Formula | CCl3COOH |
UN Number | 2564 |
Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
SMILES | C(=O)(C(Cl)(Cl)Cl)O |
IUPAC Name | 2,2,2-trichloroacetic acid |
Molecular Weight (g/mol) | 163.378 |
PubChem CID | 6421 |
Show More |
Safety and Handling
Product Identifier
- Trichloroacetic acid, solution 20% w/v, extra pure
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Trichloroacetic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction