missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TRIM47 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand: Abnova™ H00091107-T01.100uL
Additional Details : Weight : 0.00010kg
Specifications
Western Blot | |
TRIM47 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 44.11kDa | |
Transient overexpression cell lysate was tested with anti-TRIM47 antibody (Mfr. No. H00091107-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC017299.2 | |
pCMV-TRIM47 full-length | |
100 μL | |
Human |