missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ Trimethoprim

Product Code. 11757846
5 g, Each
Encompass_Preferred
Click to view available options
Quantity:
5 g
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Product Type Trimethoprim
For Use With (Application) Research
Quantity 5 g
compliance-icons compliance-icons
Product Identifier
  • Trimethoprim
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Serva Electrophoresis™ Trimethoprim

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.